Learn More
Synthware™ Inlet Adapters with Hose Connection
Hose connection for N2/Ar introduction or vacuum
Supplier: Synthware™ A532440
Description
Supplied with one Nylon cap and one Viton O-ringSpecifications
| Borosilicate Glass | |
| Inlet Adapter with Hose Connection | |
| 24/40 |