Learn More
Invitrogen™ SYBR GreenER™ qPCR SuperMix for ABI PRISM™ Instrument
Incorporate the latest high-performance technology to provide the most reliable gene expression data from your ABI 7300, 7000, 7700 or 7900HT instrument.
Supplier: Invitrogen™ 1176002K
Description
The SYBR™ GreenER™ qPCR SuperMix kits include a dsDNA-binding dye that produces a brighter signal and significantly less PCR inhibition, for improved qPCR performance over a broad dynamic range.
Consistent Specificity and Low-Copy Detection: The SYBR™ GreenER™ qPCR reagent system minimizes the formation of nonspecific products, including primer-dimers, for greater accuracy across a wide range of targets. The reliability of the SYBR™ GreenER™ qPCR reagent system also translates to greater sensitivity. With SYBR™ GreenER™ qPCR SuperMix, you can consistently detect fewer than 10 copies of a target in genomic DNA.
- The SYBR™ GreenER™ system is specially formulated to offer the best sensitivity, specificity, and reproducibility
- A novel dsDNA-binding dye - exhibits a brighter signal for increased sensitivity and less PCR inhibition than original SYBR™ Green I, while using the same instrument filters and settings
- An optimized buffer system - improves sensitivity and provides excellent long-term stability
- Uracil DNA glycosylase - reduces carryover contamination in qPCR
Specifications
| 2X | |
| SYBR | |
| qPCR | |
| 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System | |
| Real Time PCR SYBR Master Mix | |
| Gene Expression | |
| DNA (Genomic), cDNA |
| SYBR™ GreenER™ qPCR SuperMix for ABI PRISM™ instruments contains: 2X SYBR™ GreenER™ qPCR SuperMix premixed with ROX reference dye. Store at +4°C upon receipt. Guaranteed stable for 6 months when properly stored. |
|
| High | |
| Fast or Standard | |
| Platinum, SYBR GreenER | |
| 2000 Reactions | |
| 2000 Reactions |
For Research Use Only. Not for use in diagnostic procedures.