Learn More
Simport™ Scientific Swingsette™ Biopsy Processing/Embedding Cassettes in Quickload™ Stacks
Secure specimens during processing with Swingsette™ Biopsy Processing/Embedding Cassettes in Quickload™ Stacks.
$716.47 - $716.47
| Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
|
22038567
|
Simport™ Scientific
M5182T |
White |
Case of 2000 Each for $716.47
|
|
|||||
|
22038568
|
Simport™ Scientific
M5183T |
Pink |
Case of 2000 Each for $716.47
|
|
|||||
|
22038569
|
Simport™ Scientific
M5184T |
Green |
Case of 2000 Each for $716.47
|
|
|||||
|
22038570
|
Simport™ Scientific
M5185T |
Yellow |
Case of 2000 Each for $716.47
|
|
|||||
|
22038571
|
Simport™ Scientific
M5186T |
Blue |
Case of 2000 Each for $716.47
|
|
|||||
|
22038572
|
Simport™ Scientific
M5187T |
Peach |
Case of 2000 Each for $716.47
|
|
|||||
|
22038573
|
Simport™ Scientific
M5188T |
Tan |
Case of 2000 Each for $716.47
|
|
|||||
|
22038574
|
Simport™ Scientific
M5189T |
Gray |
Case of 2000 Each for $716.47
|
|
|||||
|
22038575
|
Simport™ Scientific
M51810T |
Lilac |
Case of 2000 Each for $716.47
|
|
|||||
|
22038576
|
Simport™ Scientific
M51811T |
Orange |
Case of 2000 Each for $716.47
|
|
|||||
|
22038577
|
Simport™ Scientific
M51812T |
Aqua |
Case of 2000 Each for $716.47
|
|
|||||
Description
- In Quickload™ stacks
- 45° Anterior writing area
- Easy to assemble hinged lids
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes, lids packaged separately.
Certifications
CE-IVD
Specifications
| 2000/Pk. | |
| Acetal |