Learn More
Simport™ Scientific Swingsette™ Biopsy Processing/Embedding Cassettes in Quickload™ Stacks
Secure specimens during processing with Swingsette™ Biopsy Processing/Embedding Cassettes in Quickload™ Stacks.
Supplier: Simport™ Scientific M51810T
Description
- In Quickload™ stacks
- 45° Anterior writing area
- Easy to assemble hinged lids
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes, lids packaged separately.
Certifications
CE-IVD
Specifications
| 2000/Pk. | |
| Lilac | |
| Acetal |