Learn More
Simport™ Scientific Swingsette™ Biopsy Processing/Embedding Cassettes in Quickload™ Stacks
Secure specimens during processing with Swingsette™ Biopsy Processing/Embedding Cassettes in Quickload™ Stacks.
Supplier:  Simport™ Scientific M5182T
 View more versions of this product
Catalog No. 22038567
Only null left
Explore available promotions
Description
Description
  • In Quickload™ stacks
  • 45° Anterior writing area
  • Easy to assemble hinged lids
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes, lids packaged separately.
Certifications
CE-IVD
Specifications
Specifications
2000/Pk.
White
Acetal
Product Suggestions
Product Suggestions