missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SureTect™ Beverage Spoilage Multiplex PCR Assay

Catalog No. A57289
Change view
Click to view available options
Quantity:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A57289 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A57289 Supplier Thermo Scientific™ Supplier No. A57289

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rapid qPCR solutions for beverage manufacturers. Rapidly detect a broad range of beverage spoilers from one PCR reaction with this assay.

A rapid solution to rapidly detect and differentiate yeast and mold, Brettanomyces spp., preservative-resistant yeast, lactic acid bacteria, and acetic acid bacteria in aseptically produced and carbonated product samples. Developed in partnership with a major global beverage manufacturer to meet the industry’s needs.

The SureTect™ Beverage Spoilage Multiplex PCR Assay is an optimized solution combining speed and performance in an easy-to-use and cost-effective platform, giving robust results that you can always trust.

Simple

  • Easy-to-use, simple workflow
  • Pre-dispensed lysis reagents and lyophilized PCR reagents
  • Automatic lysis cycle with very few pipetting steps

 

Fast and flexible
  • Option to automate the lysis and PCR sample preparation
  • Fast screening and differentiation of yeast and mold in a single PCR test
  • Rapid detection for early actions and high confidence in production
  • Test up to 96 samples in each run
  • Open PCR platforms

 

Smart

  • Highly specific multiplex qPCR to reduce cost and hands-on time, providing results from the four most important spoilage microorganism families for beverage, including over 100 strains, in one PCR reaction:
    • Yeast and molds (Y&M)
    • Lactic acid bacteria (LAB) and acetic acid bacteria (AAB)
    • Brettanomyces species
    • Preservative-resistant yeasts (PRY)
  • Actionable results up to 4 days earlier (depending on your current workflow) leading to risk-based decisions
  • Optimized workflow with pierceable seals to reduce cross-contamination
  • Intuitive, user-friendly software delivers reliable results, with no subjective interpretation
  • Actionable data supports fast decisions in a complex product supply chain
  • Internal positive control (IPC) in every assay for complete confidence in results

 

The SureTect™ Beverage Spoilage Multiplex PCR Assay includes:

  • Lysis Plus Tubes
  • Lysis Supplement
  • Beverage Spoilage Multiplex PCR Assay Tubes
  • Thermo Scientific 96-well Pierceable Seals (3 sheets per kit)
  • SureTect PCR Caps

 

The SureTect™ Beverage Spoilage Multiplex PCR Assay is validated on the Applied Biosystems™ QuantStudio™ 5 Food Safety Real-Time PCR System with RapidFinder™ Analysis Software v3.0 or later, allowing you to test easily and confidently. After completing the run, interpreted results are clearly displayed and can be reported, stored, printed, or downloaded as required.

Specifications

Content And Storage Store the unopened kits at 2–8°C. Store the individual components as indicated in the instructions for use.
Control Template Internal Positive Control (IPC)
Detection Method qPCR
Form Lyophilized
Format Kit, PCR Tube(s), Lysis Tube(s)
PCR Method Multiplex qPCR
Technique qPCR
For Use With (Equipment) Real-Time PCR (qPCR) System
High-throughput Compatibility High-throughput Compatible, Automated Protocols
Includes Label or Dye Yes
Kit Type QPCR Microbial Detection Kit
No. of Tests 96 Test
Product Line SureTect
Product Type PCR Assay Kit
Quantity 96 tests
Research Category Food Microbiology
Sufficient For 96 Tests
Target Organism Class Yeast, Mold, Brettanomyces spp., Lactic Acid Bacteria, Acetic Acid Bacteria
For Use With (Application) Real Time PCR (qPCR)
Compatibility RT- PCR System
Reaction Format Kit
Reagent Type PCR Assay Reagents
Sample Type Aseptically Produced Beverage Samples, Carbonated Beverage Samples, Food Samples, Environmental Samples
Starting Material Food Samples or Production Environment Samples
Volume 180 μL
Includes Lysis Plus Tubes, Lysis Supplement, Beverage Spoilage Multiplex PCR Assay Tubes, Thermo Scientific™ 96-Well Pierceable Seals (3 sheets), SureTect™ PCR Caps
Show More Show Less

For Testing of Food and Environmental Samples Only