missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ SureOne™ Large Orifice Non-Filtered Pipette Tips

Using Fisherbrand™ Sure One™ large orifice pipette tips can help maintain the integrity of genomic DNA (gDNA) and reduce the risk of cell shearing during pipetting. With a 1.5 mm inner diameter tip orifice helps reduce shearing forces and are best suited for handling viscous or particulate solutions.

$164.32 - $276.11

Specifications

Filtered Non-filtered
Color Clear
Tip Style Genomic/Wide Orifice
Product Type Non-Filtered Pipette Tip
For Use With (Equipment) Universal Fit, fits most pipettes
View More Specs

Products 5
Catalog Number Mfr. No. Volume (Metric) Sterility Format Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Sterility Format Price Quantity  
02-707-459
View Documents
N/A 100 to 1000 μL Non-sterile Hinged Rack
Pack of 960 Each for $249.86
Case of 5 Pack for $1,249.30
Only null left
 
02-707-460
View Documents
N/A 100 to 1000 μL Sterile Hinged Rack Sterile
Pack of 960 Each for $276.11
Case of 5 Pack for $1,380.55
Only null left
 
02-707-463
View Documents
Fisherbrand™
T118RFIS
1 to 200 μL Non-sterile Hinged Rack
Pack of 960 Each for $190.42
Case of 5 Pack for $952.10
Only null left
 
02-707-464
View Documents
Fisherbrand™
T118RSFIS
1 to 200 μL Sterile Hinged Rack Sterile
Pack of 960 Each for $212.26
Case of 5 Pack for $1,061.30
Only null left
 
02-707-458
View Documents
N/A 100 to 1000 μL Non-sterile Bulk
Pack of 1000 Each for $164.32
Case of 10 Pack for $1,643.20
Only null left
 
Description

Description

  • Large orifice, 1.5 mm inner diameter, helps reduce shearing forces
  • Low Retention, hydrophobic feature provides maximum sample recovery
  • Non-sterile products are certified to be free of RNase, DNase, DNA and Pyrogen
  • Sterilized products are certified to be free of RNase, DNase, DNA, Pyrogen/Endotoxin, PCR Inhibitors, ATP and Bioburden
  • Universal Fit, compatible with most popular brands of pipettor (optimal fit with Fisherbrand Elite™ pipettor)

Available bulk, rack non-sterile, rack sterile, and filtered rack sterile

Specifications

Specifications

Non-filtered
Genomic/Wide Orifice
Universal Fit, fits most pipettes
Clear
Non-Filtered Pipette Tip
Polypropylene
Videos
SDS
Documents

Documents

As part of our commitment to continuous product improvement, your delivery may contain products with some minor color variation, but their form, fit and function will not be affected. Any questions contact your sales representative.