missing translation for 'onlineSavingsMsg'
Learn More

Suramin hexasodium salt, 98%, Thermo Scientific Chemicals

Catalog No. AC328540500
Change view
Click to view available options
Quantity:
50 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC328540500 50 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC328540500 Supplier Thermo Scientific Chemicals Supplier No. 328540500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 129-46-4 | Molecular Formula: C51H34N6Na6O23S6 | Molecular Weight: 1429.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 129-46-4
Molecular Formula C51H34N6Na6O23S6
Molecular Weight (g/mol) 1429.15
MDL Number MFCD00210217
InChI Key VAPNKLKDKUDFHK-UHFFFAOYSA-H
Synonym suramin sodium, suramin hexasodium, suramin hexasodium salt, suramin sodium salt, antrypol, moranyl, naganinum, naphuride sodium, fourneau 309, suramine sodium salt
PubChem CID 8514
IUPAC Name hexasodium;8-[[4-methyl-3-[[3-[[3-[[2-methyl-5-[(4,6,8-trisulfonatonaphthalen-1-yl)carbamoyl]phenyl]carbamoyl]phenyl]carbamoylamino]benzoyl]amino]benzoyl]amino]naphthalene-1,3,5-trisulfonate
SMILES [Na+].[Na+].[Na+].[Na+].[Na+].[Na+].CC1=CC=C(C=C1NC(=O)C1=CC(NC(=O)NC2=CC(=CC=C2)C(=O)NC2=CC(=CC=C2C)C(=O)NC2=C3C(C=C(C=C3S([O-])(=O)=O)S([O-])(=O)=O)=C(C=C2)S([O-])(=O)=O)=CC=C1)C(=O)NC1=C2C(C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O)=C(C=C1)S([O-])(=O)=O

Specifications

Color White
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 50 mg
Merck Index 15, 9135
Solubility Information Solubility in water: soluble in water
Formula Weight 1429.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Suramin hexasodium salt
Show More Show Less
EINECSNumber 204-949-3

RUO – Research Use Only