missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ SupraGLN™ 100X
SupraGLN™, L-alanyl-L-glutamine dipeptide, (pronounced soo-pra gloo-tuh-meen), is a convenient alternative to L-glutamine, which is a key amino acid utilized in mammalian cell culture media.
$55.56 - $389.77
Specifications
| Concentration | 1X, 2 mM, 430 μg/mL |
|---|---|
| Sterility | Sterile |
| Formulation | 43.44 g/L of AlaGln in 0.85% Sodium Chloride |
| pH | 4.7 to 6.0 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
MP091680348
|
MP Biomedicals™
091680348 |
1 x 50 mL |
Each for $55.56
|
|
|||||
|
MP091680349
|
MP Biomedicals™
091680349 |
2 x 50 mL |
Each for $92.60
|
|
|||||
|
MP091680354
|
MP Biomedicals™
091680354 |
1 x 500 mL |
Each for $389.77
|
|
|||||
Description
- Increase stability
- Reduce ammonia build up
- Preferred glutamine source for primary cell cultures, stem cells, and high metabolic cells
Specifications
| 1X, 2 mM, 430 μg/mL | |
| Sterile | |
| 43.44 g/L of AlaGln in 0.85% Sodium Chloride | |
| 4.7 to 6.0 |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.