missing translation for 'onlineSavingsMsg'
Learn More

GVS Supported Cellulose Acetate Membranes

Catalog No. A02SP02500
Change view
Click to view available options
Pore Size:
0.2 μm
5 μm
Diameter (Metric):
13 mm
25 mm
47 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Diameter (Metric)
A02SP02500 0.2 μm 25 mm
A50SP04700 5 μm 47 mm
A02SP01300 0.2 μm 13 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A02SP02500 Supplier GVS Supplier No. 1213124
Only null left

Pure cellulose acetate impregnated on nonwoven support

  • Exceptionally strong, Triton™-free, low protein-binding cellulose acetate impregnated onto nonwoven support
  • Hydrophilic with unequaled dimensional and wet strength — eliminate filter cracking and tearing under pressure
  • Fast flow, high throughput, low protein/nucleic acid binding
  • Withstand temperatures to 180°C (356°F); autoclavable
  • Mean thickness: 150μm
  • White with plain surface; come in dispenser packs of 25 or 100

Protein and enzyme filtrations, tissue culture media preparation, biological/pharmaceutical solution filtration, sterility tests.

  1. In mL/min./cm2, measured at 520mmHg (10psi), 20°C (68°F).
  2. Pressure at which air is first forced through the pores of a water-wet membrane.
  3. Manufactured by GVS Maine

Specifications

Autoclavable Autoclavable
Bubble Point 50psi
Color White
Material Cellulose Acetate
Pore Size 0.2 μm
Flow Rate 20.0mL/min./cm sq.
For Use With (Application) Protein and enzyme filtrations; tissue culture media preparation; biological/pharmaceutical solution filtration; sterility tests.
Hydrophobicity Hydrophilic
Thickness (Metric) 150 μm
Diameter (Metric) 25 mm