missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ III First-Strand Synthesis SuperMix for qRT-PCR

Catalog No. p-4926553
Encompass
Change view
Click to view available options
No. of Reactions:
250 Reactions
50 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
11752050 50 Reactions
11752250 250 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11752050 Supplier Invitrogen™ Supplier No. 11752050
Only null left

SuperScript III First-Strand Synthesis SuperMix for qRT-PCR provides the high-temperature capability of SuperScript III Reverse Transcriptase in an optimized SuperMix format for the synthesis of first-strand cDNA for use in real-time quantitative RT-PCR (qRT-PCR).

SuperScript III First-Strand Synthesis SuperMix for qRT-PCR provides the high-temperature capability of SuperScript III Reverse Transcriptase in an optimized SuperMix format for the synthesis of first-strand cDNA for use in real-time quantitative RT-PCR (qRT-PCR). The simple, time-saving reaction set-up uses just two tubes: a 2X reaction mix and an enzyme mix.

Enzyme mix
SuperScript III Reverse Transcriptase, included in the RT enzyme mix, is a version of M-MLV RT that has been engineered to reduce RNase H activity and provide increased thermal stability. The enzyme can be used to synthesize cDNA at a temperature range of 42–60°C, providing increased specificity, higher yields of cDNA, and more full-length product than other reverse transcriptases. Because SuperScript III RT is not significantly inhibited by ribosomal and transfer RNA, it can be used to synthesize cDNA from total RNA. RNaseOUT Recombinant Ribonuclease Inhibitor, also included in the enzyme mix, is an RNase inhibitor protein that safeguards against the degradation of target RNA due to ribonuclease contamination of the RNA preparation.

Reaction mix
The 2X RT reaction mix includes oligo(dT)20, random hexamers, MgCl2, and dNTPs in a buffer formulation that has been optimized for qRT-PCR. E. coli RNase H is provided as a separate tube in the kit to remove the RNA template from the cDNA:RNA hybrid molecule after first-strand synthesis. This has been shown to increase sensitivity in qRT-PCR.

Using SuperScript III First-Strand Synthesis SuperMix
This SuperMix formulation can be used to quantify fewer than 10 copies of a target gene in qRT-PCR, with a broad dynamic range that supports accurate quantification of high-copy mRNA from up to 1 μg of total RNA. Reagents are provided for 50 or 250 RT reactions of 20 μL each.

Order Info

Shipping Condition: Dry ice

Specifications

Content And Storage

• 2X RT Reaction Mix (500 μL)
• RT Enzyme Mix (100 μL)
E. coli RNase (50 μL)

Store at –20°C.

Detection Method qPCR
Format Super Mix
GC-Rich PCR Performance High
Reaction Speed 30 min.
Technique Reverse Transcription
Optimal Reaction Temperature 50°C
Quantity 50 rxns
Reverse Transcriptase SuperScript III
Shipping Condition Dry Ice
For Use With (Application) Real Time PCR (qPCR)
Final Product Type First-Strand cDNA
No. of Reactions 50 Reactions
Reaction Format Master Mix
Reagent Type Reverse Transcription
Starting Material RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.