missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ III CellsDirect™ cDNA Synthesis Kit

Catalog No. p-4926552
Encompass
Change view
Click to view available options
No. of Reactions:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
18080300 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18080300 Supplier Invitrogen™ Supplier No. 18080300

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The SuperScript III CellsDirect cDNA Synthesis Kit is optimized for synthesis of first-strand cDNA directly from a mammalian cell lysate without first isolating the RNA.

The SuperScript III CellsDirect cDNA Synthesis Kit is optimized for synthesis of first-strand cDNA directly from a mammalian cell lysate without first isolating the RNA. Lysis and reverse transcription are performed in the same tube, and the resulting first-strand cDNA is ready to use in cloning and PCR.

Features of the SuperScript III CellsDirect Synthesis Kit include:
• Compatible with a wide range of mammalian cell types grown under different treatment conditions
• Single-tube format minimizes reagent loss, sample loss, and handling time
• Total lysate volume is used in first-strand cDNA synthesis reaction, providing greater yields with a limited number of cells and allowing for detection of rare transcripts
• SuperScript III Reverse Transcriptase, with reduced RNase H activity and higher thermal stability, produces high yields of cDNA in the first-strand synthesis reaction, for greater sensitivity and enhanced detection of rare transcripts
• Generates high-quality cDNA for use in a variety of applications, including cloning and PCR
• Simple protocol takes less than two hours

How it works
In traditional RT-PCR, RNA is first isolated from cells in a time-consuming procedure that can lead to a loss of material. Using the SuperScript III CellsDirect cDNA Synthesis Kit, the cells are lysed and the cDNA is generated from the lysate in a single tube with minimal handling and no sample loss. DNase I is added to eliminate genomic DNA prior to first-strand synthesis. This kit has been optimized for small cell samples, ranging from 10,000 cells down to a single cell (as measured by serial dilution). The use of SuperScript III Reverse Transcriptase ensures high specificity and high yields of cDNA from small amounts of starting material (as little as 10 pg total RNA). After synthesis, the first-strand cDNA can be amplified with specific primers by PCR without intermediate organic extractions or ethanol precipitations.

Specifications

Content And Storage

• Resuspension Buffer, 1 mL
• RNaseOUT™ Recombinant Ribonuclease Inhibitor, 200 μL (40 units/μL)
• DNase I, 500 μL (1 U/μL)
• 10X DNase I Buffer, 160 μL
• 25 mM EDTA, 120 μL
• Oligo(dT)20, 120 μL (50 μM)
• 10 mM dNTP Mix, 100 μL
• SuperScript™ III RT, 100 μL (200 units/μL)
• 5X RT Buffer, 600 μL
• DTT, 100 μL (0.1 M)
E. coli RNase H, 100 μL (2 U/μL)
• HeLa Total RNA, 10 μL (10 ng/μL)
• Forward/Reverse Control Primers, 10 μL (10 μM)
E. coli RNase H (2 U/μL)

Store at –20°C.

Format Kit
GC-Rich PCR Performance High
Reaction Speed 50 min.
Technique Reverse Transcription
Optimal Reaction Temperature 50°C
Quantity 100 rxns
Reverse Transcriptase SuperScript III
Ribonuclease H Activity Reduced
For Use With (Application) Real Time PCR (qPCR)
Final Product Type First-Strand cDNA
No. of Reactions 100 Reactions
Reaction Format Separate components
Reagent Type Reverse Transcription
Size (Final Product) Up to 4.5 kb
Starting Material Cells
Show More Show Less
How can I remove genomic DNA contamination from my sample prior to performing RT-PCR?

We recommend using ezDNase (Cat. No. 11766051). ezDNase Enzyme's high specificity for double-stranded DNA enables efficient and fast genomic DNA removal without reduction in the quality or quantity of RNA. ezDNase Enzyme is heat-labile and so can be easily deactivated by heat treatment at moderate temperature (55 degrees C). These features make ezDNase Enzyme an excellent choice for genomic DNA removal prior to reverse transcription reactions.

How much RNA should be employed for first-strand cDNA synthesis?

The amount of RNA template for a cDNA synthesis is highly flexible and depends upon the amount of sample available and an individual's need. In general, 1 µg total RNA is used in a typical 20-µL RT reaction.

Should I treat the cDNA with RNase H prior to downstream processing?

RNase H treatment is not always necessary. Many PCR reactions work without it. However, for cDNA synthesized with RNase H-deficient reverse transcriptases (like SuperScript II, III, and IV), RNA/cDNA hybrids—especially GC-rich ones—may not denature well, reducing PCR sensitivity. RNase H treatment can help in such cases. Additionally, RNase H treatment is beneficial for cloning larger fragments.

What percentage of RNA is converted to cDNA when performing reverse transcription?

This depends highly on the quality of the sample. mRNA itself makes up 1-5% of total RNA. Depending on the primer and enzyme used, reverse transcription can covert >70% of that into cDNA.

I'm setting up my RT reaction and am trying to decide whether I should use random primers, oligo(dT) primer, gene-specific primer, or oligo(dT)/random mix primers. What would you suggest?

Random primers are the best choice for degraded RNA, RNA with heavy secondary structure, non-polyadenylated RNA, or prokaryotic RNA. It is recommended only for two-step RT-PCR, and typically gives the highest yields, although the cDNA may not necessarily be full length. Oligo(dT) primers are good to use when trying to recover full-length cDNA from 2-step RT-PCR. The reaction is influenced by secondary structure and RNA quality. Gene specific primers should be used for very specific, mainly one-step RT-PCR reactions.

The DTT in my reverse transcription kit has precipitated—can I still use it?

No, the DTT will need to be replaced.

Are SuperScript II and III RTs RNase H minus?

These enzymes contain the domains of RNase H, but they have been mutated for reduced RNase H activity. In RNase H activity detection assays, we are not able to detect any RNase H activity.

Can I purchase the SuperScript III buffer separately?

Yes, we sell a M-MLV RT buffer (Cat. No. 18057018), which works with M-MLV RT, SuperScript II RT, and SuperScript III RT.

Will adding EDTA prior to heat-inactivation of DNase I inhibit reverse

EDTA chelates Mg2+ molecules on a 1:1 molar basis. If the amount of EDTA used for DNase I inactivation does not exceed the amount of Mg2+ present in the DNase reaction buffer, the resulting RNA solution can be used directly in the RT reaction. Otherwise, the sample should be purified to remove excess EDTA. Alternatively, consider using DNase removal methods that do not rely on EDTA or heat inactivation to avoid interference. To reduce the risk of compromised cDNA synthesis, we recommend performing gDNA removal with ezDNase Enzyme (Cat. No. 11766051) which is specific to dsDNA and heat-labile, hence does not require harsh inactivation conditions.

Does SuperScript III exhibit TdT activity?

SuperScript III exhibits low TdT activity. If TdT activity is required, use our SuperScript IV RT or SuperScript IV Template Switching RT Master Mix.

What is the difference between SuperScript III RT and the RT in the SuperScript VILO kit?

The SuperScript VILO cDNA Synthesis Kit contains a mix of SuperScript III RT and helper proteins which help to increase the efficiency of the reverse transcription reaction and thus improve yield. The RT in the SuperScript VILO kit is active at 42 degrees C due to the helper proteins.

Do you have a kit for cDNA synthesis directly from mammalian cells?

Yes, we do offer the SuperScript CellsDirect cDNA Synthesis Kit which can be used for 1 cell up to 10,000 cells. This kit can also be used for frozen LCM samples but not formalin fixed/paraffin embedded tissues. Alternatively, you could also use our Ambion Cells-to-CT kits which provides RT-PCR amplification directly from cell lysates. These kits are compatible with cultured cells and LCM samples.

I want to perform qRT-PCR using cDNA that I synthesize directly from cells. Can I use the SuperScript III CellsDirect cDNA Kit for end-point PCR and the Platinum Quantitative PCR SuperMix-UDG kit?

We would recommend using our Cells-to-CT kits for best qRT-PCR results. The CellsDirect kits for end-point PCR have not been optimized for qRT-PCR.


For Research Use Only. Not for use in diagnostic procedures.