missing translation for 'onlineSavingsMsg'
Learn More

Super-DHB suitable for matrix substance for MALDI-MS, ≥99% (HPLC), MilliporeSigma™ Supelco™

$168.96 - $728.81

Specifications

CAS 63542-76-7
Purity Grade Notes 9:1 mixture of DHB and 2-hydroxy-5-methoxybenzoic acid
Solubility Soluble in methanol
Type Organic Standard
Percent Purity ≥99% (sum of DHB and 2-hydroxy-5-methoxybenzoi c acid, HPLC)
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111014995
View Documents
Merck Emd Millipore
5086210X10MGF
10 x 10 mg
Each for $490.91
Only null left
 
111014993
View Documents
Merck Emd Millipore
508621GF
1 g
Each for $168.96
Only null left
 
111014994
View Documents
Merck Emd Millipore
508625GF
5 g
Each for $728.81
Only null left
 
Description

Description

The “super-DHB” matrix consists of a 9:1 (w/w) mixture of 2,5-DHB and 2-hydroxy-5-methoxybenzoic acid, respectively, since this ratio was found to be the most effective. The beneficial effect of the 2-hydroxy- 5-methoxybenzoic acid additive is attributed to a disorder in the 2,5-DHB crystal lattice resulting in enhanced ion desorption (i.e. ‘softer’ desorption) and reduced internal energy, which in turn results in reduced metastable fragmentation. An increase in sensitivity of up to 2 to 3-fold has been reported for a standard dextran mixture with concomitant enhanced resolution attributed to reduced metastable ion formation. The super-DHB matrix has also been reported to generate enhanced signals from tryptic glycopeptides. The use of super-DHB matrix is reported to reduce the presence of matrix background ions and result in glycan signals exhibiting better signal to noise ratios and higher resolution in the mass spectrum.
Specifications

Specifications

63542-76-7
Soluble in methanol
≥99% (sum of DHB and 2-hydroxy-5-methoxybenzoi c acid, HPLC)
9:1 mixture of DHB and 2-hydroxy-5-methoxybenzoic acid
Organic Standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'