missing translation for 'onlineSavingsMsg'
Learn More

Super-DHB suitable for matrix substance for MALDI-MS, ≥99% (HPLC), MilliporeSigma™ Supelco™

Supplier:  Merck Emd Millipore 508621GF

 View more versions of this product

Catalog No. 111014993


Only null left
Explore available promotions

Description

Description

The “super-DHB” matrix consists of a 9:1 (w/w) mixture of 2,5-DHB and 2-hydroxy-5-methoxybenzoic acid, respectively, since this ratio was found to be the most effective. The beneficial effect of the 2-hydroxy- 5-methoxybenzoic acid additive is attributed to a disorder in the 2,5-DHB crystal lattice resulting in enhanced ion desorption (i.e. ‘softer’ desorption) and reduced internal energy, which in turn results in reduced metastable fragmentation. An increase in sensitivity of up to 2 to 3-fold has been reported for a standard dextran mixture with concomitant enhanced resolution attributed to reduced metastable ion formation. The super-DHB matrix has also been reported to generate enhanced signals from tryptic glycopeptides. The use of super-DHB matrix is reported to reduce the presence of matrix background ions and result in glycan signals exhibiting better signal to noise ratios and higher resolution in the mass spectrum.
Specifications

Specifications

63542-76-7
Soluble in methanol
1 g
9:1 mixture of DHB and 2-hydroxy-5-methoxybenzoic acid
Organic Standard
≥99% (sum of DHB and 2-hydroxy-5-methoxybenzoi c acid, HPLC)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions