missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Super AquaBlue ELISA Substrate

Catalog No. 5018493
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5018493 500 mL
501122023 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5018493 Supplier Invitrogen™ Supplier No. 00420358
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Enhanced ABTS substrate solution for ELISA which provides high signal and sensitivity, low background. Safe, stable and convenient to use. Oxidized by HRP-labeled probes, resulting in soluble, blue-green colored end product, whose absorption can be measured by spectrophotometers at 405nm.

  • Highly sensitive and extremely stable (18 months at room temperature or 24 months at 4°C).
  • Engineered for convenience as a single component (no additional H2O2 or other components are necessary)
  • Suitable for endpoint reactions, but is also ideal for kinetic assays; stopping the reaction with acid stop solution does not change the color or absorption properties of the blue-green end product

Specifications

Product Type ELISA Substrate
Quantity 500 mL
Detection Method Colorimetric
Substrate Properties Chemical Substrate
Substrate Type HRP (Horseradish Peroxidase)

For Research Use Only. Not for use in diagnostic procedures.