missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Supelguard™ Guard Cartridge Holder - Direct-Connect

Catalog No. 111010831
Change view
Click to view available options
For use with Supelguard cartridge (2 cm L. x 3 to 4.6 mm I.D.)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111010831 For use with Supelguard cartridge (2 cm L. x 3 to 4.6 mm I.D.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111010831 Supplier MilliporeSigma™ Supelco™ Supplier No. 55205

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with Supelguard cartridge (2 cm L. x 3 to 4.6 mm I.D.)

Connects guard column directly to a modular analytical column.

Specifications

Length (Metric) 2 cm
Type Guard Cartridge Holder
For Use With (Equipment) Supelguard Cartridge (2 cm L. x 3 to 4.6 mm I.D.)
Material Stainless Steel Housing
For Use With (Application) HPLC
Product Line Supelguard

Supelguard is a trademark of Sigma-Aldrich Co. LLC