missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ SUPELCOSIL™ LC-Diol HPLC Column, 5 μm Particle Size

LC-Diol columns can be used to separate proteins by gel filtration chromatography.

$1650.55 - $1697.71

Specifications

Length (Metric) 25 cm
Particle Size 5 μm
Pore Size 120 Å
Surface Area 170 m2/g
Stationary Phase Diol
View More Specs

Products 2
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
111003201
View Documents
MilliporeSigma™ Supelco™
58201C40
4 mm
Each for $1,650.55
Only null left
 
111003199
View Documents
MilliporeSigma™ Supelco™
SGMA58201
4.6 mm
Each for $1,697.71
Only null left
 
Description

Description

When operated with an aqueous buffer, the diol phase can effectively shield the silica surface from interacting with proteins. A well-known use of diol columns, under normal phase conditions, is the separation of steroids and sterols.
Specifications

Specifications

25 cm
120 Å
Diol
Diol Phase/Silica Gel, Spherical Particle Platform
Reversed Phase
5 μm
170 m2/g
SUPELCOSIL
No
Videos
SDS
Documents

Documents

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC