missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ SUPELCOGEL™ 8H, 8% Crosslinked HPLC Column, 9 μm Particle Size

Catalog No. 111006257
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111006257 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111006257 Supplier MilliporeSigma™ Supelco™ Supplier No. 59246U

Offers excellent separation of many common samples.

SUPELCOGEL™ 8 Column Series (manufactured with 8% XL PS-DVB):The majority of columns on the market for the analysis of carbohydrates, organic acids, and alcohols, such as Aminex™, utilize an 8% XL PS-DVB. The 8% polymer offers excellent separation of many common samples. Advantages: 8% polymers combine durability and good resolution. Due to higher cross-linking than 6% XL polymers, columns packed with 8% polymer can be used at higher flow rates therefore decreasing analysis times.Disadvantages: 8% polymers are limited in separating carbohydrates larger than trisaccharides due to lower MW exclusion limits. Compared to the 6% XL polymer, for some types of carbohydrate and organic acid mixtures, the 8% shows less resolution due to selectivity differences. Molecular Weight Exclusion Limit: 1200 minimum. Wet Capacity meq/mL: 1.7 - 1.9. Flow Rate Recommendation: 0.4 - 1.2 ml/min.

Specifications

Crosslinking 0.08
Diameter (Metric) 7.8 mm
Length (Metric) 30 cm
Particle Size 9 μm
Stationary Phase Sulfonated Polystyrene
Product Line SUPELCOGEL
Matrix Sulfonated Polystyrene/Divinylbenzene Phase/Spherical Particle Platform
Phase Ion Exclusion
Technique Crosslinked HPLC

SUPELCOGEL is a trademark of Sigma-Aldrich Co. LLC