missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ SUPELCOGEL™ 8Ca, 8% Crosslinked HPLC Column, 9 μm Particle Size
Description
SUPELCOGEL™ 8 Column Series (manufactured with 8% XL PS-DVB): The majority of columns on the market for the analysis of carbohydrates, organic acids, and alcohols, such as Aminex™, utilize an 8% XL PS-DVB. The 8% polymer offers excellent separation of many common samples. Advantages: 8% polymers combine durability and good resolution. Due to higher cross-linking than 6% XL polymers, columns packed with 8% polymer can be used at higher flow rates therefore decreasing analysis times. Disadvantages: 8% polymers are limited in separating carbohydrates larger than trisaccharides due to lower MW exclusion limits. Compared to the 6% XL polymer, for some types of carbohydrate and organic acid mixtures, the 8% shows less resolution due to selectivity differences. Molecular Weight Exclusion Limit: 1200 minimum Wet Capacity meq/mL: 1.7 - 1.9 Flow Rate Recommendation: 0.4 - 1.2 mL/min
Specifications
Specifications
| Crosslinking | 0.08 |
| Diameter (Metric) | 4.6 mm |
| Length (Metric) | 25 cm |
| Particle Size | 9 μm |
| Stationary Phase | Sulfonated Polystyrene |
| Product Line | SUPELCOGEL |
| Matrix | Sulfonated Polystyrene/Divinylbenzene Phase/Spherical Particle Platform |
| Phase | Ion Exclusion |
| Technique | Crosslinked HPLC |
SUPELCOGEL is a trademark of Sigma-Aldrich Co. LLC