missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ SUN-SRi™ Crimp Caps
Thermo Scientific caps and vials are available in a wide variety of sizes and configurations to fit the needs of many applications.
Supplier: Thermo Scientific™ 502370
Description
Thermo Scientific caps and vials are available in a wide variety of sizes and configurations to fit the needs of many applications.
- Tin plate caps or two-part composite tin plate and aluminum cap with or without septum
- Two-part caps feature aluminum skirts that make the closures easier to crimp and remove
- 20mm Tin Plate
Specifications
| Without Septum | |
| Aluminum | |
| 500/Pk. | |
| 20 mm |
| Blue | |
| Magnetic ring | |
| Crimp Cap | |
| Headspace Vials |