missing translation for 'onlineSavingsMsg'
Learn More

Sultamicillin Tosylate Dihydrate 98.0+%, TCI America™

$169.63 - $467.87

Chemical Identifiers

CAS 83105-70-8
Molecular Formula C32H42N4O14S3
Molecular Weight (g/mol) 802.882
MDL Number MFCD01682151
InChI Key KNVDAMRBJYZXRW-ZLNTYAKYSA-N
Synonym Sultamicillin Tosilate
PubChem CID 71775005
IUPAC Name [(5R)-3,3-dimethyl-4,4,7-trioxo-4$l^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carbonyl]oxymethyl (2S,5R,6R)-6-[[(2S)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;4-methylbenzenesulfonic acid;dihydrate
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)OCOC(=O)C4C(S(=O)(=O)C5N4C(=O)C5)(C)C)C.O.O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
U0097250MG
View Documents
TCI America
U0097250MG
250 mg
Each for $169.63
Only null left
 
U00971G
View Documents
TCI America
U00971G
1 g
Each for $467.87
Only null left
 
Description

Description

Specifications

Chemical Identifiers

83105-70-8
802.882
KNVDAMRBJYZXRW-ZLNTYAKYSA-N
71775005
CC1=CC=C(C=C1)S(=O)(=O)O.CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)OCOC(=O)C4C(S(=O)(=O)C5N4C(=O)C5)(C)C)C.O.O
C32H42N4O14S3
MFCD01682151
Sultamicillin Tosilate
[(5R)-3,3-dimethyl-4,4,7-trioxo-4$l^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carbonyl]oxymethyl (2S,5R,6R)-6-[[(2S)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;4-methylbenzenesulfonic acid;dihydrate

Specifications

83105-70-8
C32H42N4O14S3
250 mg
KNVDAMRBJYZXRW-ZLNTYAKYSA-N
[(5R)-3,3-dimethyl-4,4,7-trioxo-4$l^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carbonyl]oxymethyl (2S,5R,6R)-6-[[(2S)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;4-methylbenzenesulfonic acid;dihydrate
71775005
≥98.0% (GC,T)
Sultamicillin Tosylate Dihydrate
White-Yellow
MFCD01682151
Sultamicillin Tosilate
CC1=CC=C(C=C1)S(=O)(=O)O.CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)OCOC(=O)C4C(S(=O)(=O)C5N4C(=O)C5)(C)C)C.O.O
802.882
766.85
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : XH8460000

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents