missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sultamicillin Tosylate Dihydrate 98.0+%, TCI America™
Supplier: TCI America U0097250MG
Specifications
| Sultamicillin Tosylate Dihydrate | |
| White-Yellow | |
| MFCD01682151 | |
| Sultamicillin Tosilate | |
| CC1=CC=C(C=C1)S(=O)(=O)O.CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)OCOC(=O)C4C(S(=O)(=O)C5N4C(=O)C5)(C)C)C.O.O | |
| 802.882 | |
| 766.85 | |
| Crystalline Powder |
| 83105-70-8 | |
| C32H42N4O14S3 | |
| 250 mg | |
| KNVDAMRBJYZXRW-ZLNTYAKYSA-N | |
| [(5R)-3,3-dimethyl-4,4,7-trioxo-4$l^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carbonyl]oxymethyl (2S,5R,6R)-6-[[(2S)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;4-methylbenzenesulfonic acid;dihydrate | |
| 71775005 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 83105-70-8 | |
| 802.882 | |
| KNVDAMRBJYZXRW-ZLNTYAKYSA-N | |
| 71775005 | |
| CC1=CC=C(C=C1)S(=O)(=O)O.CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)OCOC(=O)C4C(S(=O)(=O)C5N4C(=O)C5)(C)C)C.O.O |
| C32H42N4O14S3 | |
| MFCD01682151 | |
| Sultamicillin Tosilate | |
| [(5R)-3,3-dimethyl-4,4,7-trioxo-4$l^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carbonyl]oxymethyl (2S,5R,6R)-6-[[(2S)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;4-methylbenzenesulfonic acid;dihydrate |
Safety and Handling
RTECSNumber : XH8460000
TSCA : No
Recommended Storage : Refrigerator