missing translation for 'onlineSavingsMsg'
Learn More

Sultamicillin Tosylate Dihydrate 98.0+%, TCI America™

Supplier:  TCI America U00971G

 View more versions of this product

Catalog No. U00971G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sultamicillin Tosylate Dihydrate
White-Yellow
MFCD01682151
Sultamicillin Tosilate
CC1=CC=C(C=C1)S(=O)(=O)O.CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)OCOC(=O)C4C(S(=O)(=O)C5N4C(=O)C5)(C)C)C.O.O
802.882
766.85
Crystalline Powder
83105-70-8
C32H42N4O14S3
1 g
KNVDAMRBJYZXRW-ZLNTYAKYSA-N
[(5R)-3,3-dimethyl-4,4,7-trioxo-4$l^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carbonyl]oxymethyl (2S,5R,6R)-6-[[(2S)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;4-methylbenzenesulfonic acid;dihydrate
71775005
≥98.0% (GC,T)

Chemical Identifiers

83105-70-8
802.882
KNVDAMRBJYZXRW-ZLNTYAKYSA-N
71775005
CC1=CC=C(C=C1)S(=O)(=O)O.CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)OCOC(=O)C4C(S(=O)(=O)C5N4C(=O)C5)(C)C)C.O.O
C32H42N4O14S3
MFCD01682151
Sultamicillin Tosilate
[(5R)-3,3-dimethyl-4,4,7-trioxo-4$l^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carbonyl]oxymethyl (2S,5R,6R)-6-[[(2S)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;4-methylbenzenesulfonic acid;dihydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : XH8460000

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions