missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Sulpiride 98.0+%, TCI America™

Supplier:  TCI America S050125G

 View more versions of this product

Catalog No. S050125G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(+/-)-Sulpiride
185°C
C15H23N3O4S
25 g
BGRJTUBHPOOWDU-UHFFFAOYNA-N
N-[(1-ethylpyrrolidin-2-yl)methyl]-2-methoxy-5-sulfamoylbenzamide
5355
341.43
Crystalline Powder
15676-16-1
White
MFCD00055061
sulpiride, sulpyrid, sulpirid, aiglonyl, dolmatil, dogmatil, guastil, mirbanil, misulvan, sursumid
CCN1CCCC1CNC(=O)C1=CC(=CC=C1OC)S(N)(=O)=O
341.43
CHEBI:32168
≥98.0% (HPLC,T)

Chemical Identifiers

15676-16-1
341.43
BGRJTUBHPOOWDU-UHFFFAOYNA-N
5355
N-[(1-ethylpyrrolidin-2-yl)methyl]-2-methoxy-5-sulfamoylbenzamide
C15H23N3O4S
MFCD00055061
sulpiride, sulpyrid, sulpirid, aiglonyl, dolmatil, dogmatil, guastil, mirbanil, misulvan, sursumid
CHEBI:32168
CCN1CCCC1CNC(=O)C1=CC(=CC=C1OC)S(N)(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-2168

RTECSNumber : BZ3400000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions