missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Sulpiride 98.0+%, TCI America™

Catalog No. p-7135911
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S05015G 5 g
S050125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S05015G Supplier TCI America Supplier No. S05015G

Chemical Identifiers

CAS 15676-16-1
Molecular Formula C15H23N3O4S
Molecular Weight (g/mol) 341.43
MDL Number MFCD00055061
InChI Key BGRJTUBHPOOWDU-UHFFFAOYNA-N
Synonym sulpiride, sulpyrid, sulpirid, aiglonyl, dolmatil, dogmatil, guastil, mirbanil, misulvan, sursumid
PubChem CID 5355
ChEBI CHEBI:32168
IUPAC Name N-[(1-ethylpyrrolidin-2-yl)methyl]-2-methoxy-5-sulfamoylbenzamide
SMILES CCN1CCCC1CNC(=O)C1=CC(=CC=C1OC)S(N)(=O)=O

Specifications

Melting Point 185°C
Color White
Quantity 5 g
Formula Weight 341.43
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (+/-)-Sulpiride
EINECSNumber (9)-2168
RTECSNumber BZ3400000
TSCA No