missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid Solution (0.02N (N/50)/Certified), Fisher Chemical™

Catalog No. p-20639
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Poly Bottle
PolyPac™
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SA2261 1 L Poly Bottle
SA2264 4 L Poly Bottle
SA22620 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SA2261 Supplier Thermo Fisher Scientific Supplier No. FLSA2261
Only null left

Standardized to pH 4.4 at 25°C against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5 , 50-00-0
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 4.4
Color Colorless
Boiling Point 100°C
Concentration 0.0198 to 0.0202N
Identification Pass Test
Packaging Poly Bottle
Traceability to NIST LOT 84L Potassium acid phthalate
Quantity 1 L
Physical Form Liquid
UN Number 2796
Grade Certified
Concentration or Composition (by Analyte or Components) 0.02N (N/50)
Chemical Name or Material Sulfuric Acid Solution, 0.02N (N/50)
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
Contains a known or suspected carcinogen. The toxicological properties have not been fully investigated. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:0
DOTInformation DOT Class 8, : Corrosive