missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solution d’acide sulfurique (0,02N (N/50)/Certifié), Fisher Chemical™
Specifications
Specifications
| pH | 4.4 |
| Nom ou substance chimique | Sulfuric Acid Solution |
| Couleur | Incolore |
| Nom Remarque | 0.02N (N/50) |
| Concentration | 0,0198 à 0,0202N |
| Point d’ébullition | 100 °C |
| Désignation | Test Réussi |
| Emballage | Bouteille en polyéthylène |
| Traçabilité NIST | LOT 84L Phtalate d’acide potassique |
| Quantité | 4 L |
| Show More |
Safety and Handling
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview Contains a known or suspected carcinogen. The toxicological properties have not been fully investigated. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention. NFPA Health:1 Flammability:0 Instability:0 |
| DOTInformation | DOT Class 8, : Corrosive |