missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 50% (v/v), Ricca Chemical
$88.69 - $1,660.66
Chemical Identifiers
| CAS | 7664-93-9 |
|---|---|
| Molecular Formula | H2O4S |
| Molecular Weight (g/mol) | 98.07 |
| MDL Number | MFCD00064589 |
| InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
| Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
| PubChem CID | 1118 |
| ChEBI | CHEBI:26836 |
| IUPAC Name | sulfuric acid |
| SMILES | OS(O)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
8180-16
|
Ricca Chemical Company
818016 |
500 mL | Natural Poly Bottle |
N/A
|
|
||||
|
8180-32
|
Ricca Chemical Company
RI818032 |
1 L | Natural Poly Bottle |
N/A
|
|
||||
|
8180-5PT
|
Ricca Chemical Company
81805PT |
2.5 L | Safety-coated Glass Bottle |
N/A
|
|
||||
|
8180-1
|
Ricca Chemical Company
81801 |
4 L | Natural Poly Bottle |
N/A
|
|
||||
|
8180-5
|
Ricca Chemical Company
81805 |
20 L | Cubitainer |
N/A
|
|
||||
|
81805HP
|
Ricca Chemical Company
81805HP |
20 L | Hard Pack |
N/A
|
|
||||
Chemical Identifiers
| 7664-93-9 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| 1118 | |
| sulfuric acid |
| H2O4S | |
| MFCD00064589 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| CHEBI:26836 | |
| OS(O)(=O)=O |
Specifications
| 7664-93-9,7732-18-5 | |
| 51.0,51.0 | |
| Aqueous solution | |
| H2O4S | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| OS(O)(=O)=O | |
| 98.07 | |
| CHEBI:26836 | |
| Laboratory | |
| Sulfuric Acid, Approximately 18N |
| 49.0,49.0 | |
| Natural Poly Bottle | |
| 500 mL | |
| MFCD00064589 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| 50% V/V | |
| 50%(v/v) |
Safety and Handling
Recommended Storage : 15-30°C