missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 50% (v/v), Ricca Chemical
Supplier: Ricca Chemical Company 81805HP
Specifications
| Sulfuric Acid | |
| 7664-93-9,7732-18-5 | |
| 51.0,51.0 | |
| Aqueous solution | |
| H2O4S | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| OS(O)(=O)=O | |
| 98.07 | |
| CHEBI:26836 | |
| Laboratory |
| Approximately 18N | |
| 49.0,49.0 | |
| Hard Pack | |
| 20 L | |
| MFCD00064589 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| 50% V/V | |
| 50%(v/v) |
Chemical Identifiers
| 7664-93-9 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| 1118 | |
| sulfuric acid |
| H2O4S | |
| MFCD00064589 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| CHEBI:26836 | |
| OS(O)(=O)=O |
Safety and Handling
Recommended Storage : 15-30°C