missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 50% (v/v), Ricca Chemical

Supplier:  Ricca Chemical Company 81801

 View more versions of this product

Catalog No. 8180-1


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sulfuric Acid
7664-93-9,7732-18-5
51.0,51.0
Aqueous solution
H2O4S
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836
Laboratory
Approximately 18N
49.0,49.0
Natural Poly Bottle
4 L
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
50% V/V
50%(v/v)

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : 15-30°C

SDS Documents

Documents

Accessories
Promotions

Promotions