missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 10% (v/v), Ricca Chemical
$75.94 - $1644.78
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O4S |
| Molecular Weight (g/mol) | 98.07 |
| MDL Number | MFCD00064589 |
| InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
| Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
| PubChem CID | 1118 |
| ChEBI | CHEBI:26836 |
| IUPAC Name | sulfuric acid |
| SMILES | OS(O)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
8150-16
|
Ricca Chemical Company
815016 |
500 mL | Natural Poly Bottle |
Each for $75.94
|
|
||||
|
8150-32
|
Ricca Chemical Company
815032 |
1 L | Natural Poly Bottle |
Each for $117.84
|
|
||||
|
81505PT
|
Ricca Chemical Company
81505PT |
2.5 L | Safety-coated Glass Bottle |
Each for $230.25
|
|
||||
|
8150-1
|
Ricca Chemical Company
RI81501 |
4 L | Natural Poly Bottle |
Each for $306.47
|
|
||||
|
815016G
|
Ricca Chemical Company
815016G |
16 gal. | Poly Drum |
Each for $1,644.78
|
|
||||
Chemical Identifiers
| 7732-18-5 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| 1118 | |
| sulfuric acid |
| H2O4S | |
| MFCD00064589 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| CHEBI:26836 | |
| OS(O)(=O)=O |
Specifications
| 7732-18-5,7664-93-9 | |
| 91.0,11.0 | |
| Aqueous solution | |
| H2O4S | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| OS(O)(=O)=O | |
| 98.07 | |
| CHEBI:26836 | |
| Laboratory | |
| Sulfuric Acid |
| 89.0,9.0 | |
| Natural Poly Bottle | |
| 500 mL | |
| MFCD00064589 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| 10% V/V | |
| 10%(v/v) |