missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 0.500 N (N/2), Ricca Chemical
$63.10 - $494.27
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O4S |
| Molecular Weight (g/mol) | 98.07 |
| MDL Number | MFCD00064589 |
| InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
| Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
| PubChem CID | 1118 |
| ChEBI | CHEBI:26836 |
| IUPAC Name | sulfuric acid |
| SMILES | OS(O)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
828016
|
Ricca Chemical Company
828016 |
500 mL | Natural Poly Bottle |
Each for $63.10
|
|
||||
|
8280-32
|
Ricca Chemical Company
828032 |
1 L | Natural Poly Bottle |
Each for $85.67
|
|
||||
|
8280-1
|
Ricca Chemical Company
82801 |
4 L | Natural Poly Bottle |
Each for $214.73
|
|
||||
|
8280-5
|
Ricca Chemical Company
82805 |
20 L | Cubitainer |
Each for $494.27
|
|
||||
Chemical Identifiers
| 7732-18-5 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| 1118 | |
| sulfuric acid |
| H2O4S | |
| MFCD00064589 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| CHEBI:26836 | |
| OS(O)(=O)=O |
Specifications
| Colorless | |
| 96.0,2.45 | |
| 99.0,2.55 | |
| 500 mL | |
| Liquid | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| OS(O)(=O)=O | |
| 98.07 | |
| CHEBI:26836 | |
| Sulfuric Acid |
| 7732-18-5,7664-93-9 | |
| 0.500Normal | |
| Natural Poly Bottle | |
| H2O4S | |
| MFCD00064589 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| Laboratory |