missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 0.500 N (N/2), Ricca Chemical

Supplier:  Ricca Chemical Company 828032

 View more versions of this product

Catalog No. 8280-32


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sulfuric Acid
7732-18-5,7664-93-9
0.500Normal
Natural Poly Bottle
H2O4S
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
Laboratory
Colorless
96.0,2.45
99.0,2.55
1 L
Liquid
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836

Chemical Identifiers

7732-18-5
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions