missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, 0.1N Standardized Solution

Catalog No. p-4408599
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35651K2 1 L
AA35651K7 4 L
AA35651M4 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA35651K2 Supplier Thermo Scientific Chemicals Supplier No. 035651K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

Sulfuric acid, 0.1N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Density 1g/cm3 at 20°C
Physical Form Liquid
Solubility Miscible with water
Type Analytical Volumetric Solution
Quantity 1 L
Formula Weight 98.08
Grade Standard
Concentration or Composition (by Analyte or Components) Sulfuric acid: 0.05%; Water: 99.95%
Chemical Name or Material Sulfuric acid
EINECSNumber 231-639-5
RTECSNumber WS5600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only