missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid, 0.1N Standardized Solution
Description
Sulfuric acid, 0.1N standardized solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
| Density | 1g/cm3 at 20°C |
| Physical Form | Liquid |
| Solubility | Miscible with water |
| Quantity | 20 L |
| Formula Weight | 98.08 |
| Concentration or Composition (by Analyte or Components) | Sulfuric acid: 0.05%; Water: 99.95% |
| Chemical Name or Material | Sulfuric acid |
Safety and Handling
| EINECSNumber | 231-639-5 |
| RTECSNumber | WS5600000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only