missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, 0.02N Standardized Solution

Catalog No. AA35649K7
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35649K7 4 L
AA35649K2 1 L
AA35649M4 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA35649K7 Supplier Thermo Scientific Chemicals Supplier No. 035649K7

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for volumetric analysis in laboratories. | CAS: 7664-93-9 | H2O4S | 98.07 g/mol

Sulfuric acid, 0.02N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Physical Form Liquid
Solubility Miscible with water
Type Analytical Volumetric Solution
Quantity 4 L
Formula Weight 98.08
Grade Standard
Chemical Name or Material Sulfuric acid
EINECSNumber 231-639-5
RTECSNumber WS5600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only