missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Trioxide - Triethylamine Complex 96.0+%, TCI America™

$134.21 - $434.32

Chemical Identifiers

CAS 761-01-3
Molecular Formula C6H15NO3S
Molecular Weight (g/mol) 181.25
InChI Key YYHPEVZFVMVUNJ-UHFFFAOYSA-N
Synonym Triethylamine - Sulfur Trioxide Complex
PubChem CID 222301
IUPAC Name N,N-diethylethanamine;sulfur trioxide
SMILES CCN(CC)CC.O=S(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T21365G
View Documents
TCI America
T21365G
5 g N/A
Only null left
 
T213625G
View Documents
TCI America
T213625G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

761-01-3
181.25
Triethylamine - Sulfur Trioxide Complex
N,N-diethylethanamine;sulfur trioxide
C6H15NO3S
YYHPEVZFVMVUNJ-UHFFFAOYSA-N
222301
CCN(CC)CC.O=S(=O)=O

Specifications

93°C
761-01-3
5 g
3261
YYHPEVZFVMVUNJ-UHFFFAOYSA-N
N,N-diethylethanamine;sulfur trioxide
222301
≥96.0% (N)
White-Yellow
Crystalline Powder
C6H15NO3S
Triethylamine - Sulfur Trioxide Complex
CCN(CC)CC.O=S(=O)=O
181.25
181.25
Sulfur Trioxide - Triethylamine Complex
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'