missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Trioxide - Triethylamine Complex 96.0+%, TCI America™

Supplier:  TCI America T213625G

 View more versions of this product

Catalog No. T213625G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sulfur Trioxide - Triethylamine Complex
White-Yellow
Crystalline Powder
C6H15NO3S
Triethylamine - Sulfur Trioxide Complex
CCN(CC)CC.O=S(=O)=O
181.25
181.25
93°C
761-01-3
25 g
3261
YYHPEVZFVMVUNJ-UHFFFAOYSA-N
N,N-diethylethanamine;sulfur trioxide
222301
≥96.0% (N)

Chemical Identifiers

761-01-3
181.25
Triethylamine - Sulfur Trioxide Complex
N,N-diethylethanamine;sulfur trioxide
C6H15NO3S
YYHPEVZFVMVUNJ-UHFFFAOYSA-N
222301
CCN(CC)CC.O=S(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions