missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur Trioxide - Triethylamine Complex 96.0+%, TCI America™
Supplier: TCI America T21365G
Specifications
| Sulfur Trioxide - Triethylamine Complex | |
| White-Yellow | |
| Crystalline Powder | |
| C6H15NO3S | |
| Triethylamine - Sulfur Trioxide Complex | |
| CCN(CC)CC.O=S(=O)=O | |
| 181.25 | |
| 181.25 |
| 93°C | |
| 761-01-3 | |
| 5 g | |
| 3261 | |
| YYHPEVZFVMVUNJ-UHFFFAOYSA-N | |
| N,N-diethylethanamine;sulfur trioxide | |
| 222301 | |
| ≥96.0% (N) |
Chemical Identifiers
| 761-01-3 | |
| 181.25 | |
| Triethylamine - Sulfur Trioxide Complex | |
| N,N-diethylethanamine;sulfur trioxide |
| C6H15NO3S | |
| YYHPEVZFVMVUNJ-UHFFFAOYSA-N | |
| 222301 | |
| CCN(CC)CC.O=S(=O)=O |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator