missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Oil Standards for Diesel Fuel Analysis in Base Oil, Set of 8, SPEX CertiPrep™

Sulfur Oil Standard Set in Base Oil 20, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.

Supplier:  SPEX CertiPrep Group LLC DSS8SET

Catalog No. DSS8-SET


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Sulfur Oil Standard Set
  • Includes sulfur standards in Base Oil 20 in concentrations of 50, 100, 200, 300, 500, 750 and 1,000 μg/g
  • Also includes Base Oil Blank
  • 100 mL volume for each component in the set

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Sulfur Oil Standards in Base Oil
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Certified Reference Material
Petrochemical Testing
Oil
Set of 7 Standards and Base Oil 20 Blank
With Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Base Oil 20
1 Kit
Set contains one 100g bottle each:DSS8-AYDSS8-1YDSS8-BYDSS8-CYDSS8-1AYDSS8-1BYDSS8-2YSet contains one 500g bottle:BASE 20 (blank)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only