Learn More
Thermo Scientific™ SulfoLink™ Coupling Resin
Description
Thermo Scientific™ SulfoLink Coupling Resin is iodoacetyl-activated for the covalent immobilization of sulfhydryl-containing peptides or proteins for affinity purification.
Complete SulfoLink Immobilization Kits are available for peptides and proteins. Each kit contains all the reagents for preparing the respective peptide or protein sample and five columns containing 2mL of SulfoLink Resin that can be used in either gravity-flow or centrifuge format for efficient coupling reactions and multiple cycles of affinity purification. A single-column Trial Kit for use with either peptides or proteins is also available, or the SulfoLink Coupling Resin can be purchased separately.
Highlights:
Specific conjugation through sulfhydryl (-SH) groups – the iodoacetyl groups react specifically with sulhydryls to form irreversible thioether bonds
Separate kits optimized for peptides or proteins – kits include optimized reagents for preparing peptide or protein samples for efficient immobilization
Fast – spin columns increase protocol speed; prepare and couple samples in 2 hours (peptides) to 3.5 hours (proteins)
Flexible coupling conditions – use pH 7.5 to 9.0 aqueous buffers, organic solvent (e.g., 20% DMSO) or denaturant (guanidine·HCl), as needed for protein or peptide solubility during coupling reaction
Easy-to-follow instructions – streamlined protocols for sample preparation, immobilization and affinity purification
High capacity – immobilize 1 to 2mg peptide or 2 to 20mg protein per 2mL column of SulfoLink Coupling Resin
Includes:
Kits contain SulfoLink Resin Columns (2mL), protein or peptide preparation reagents (e.g., reducing agent), coupling buffer, wash solution, L-cysteine solution (for quenching) and desalting columns (protein kit only)
Recommended for:
Immobilize peptides having terminal cysteine residues to purify antibodies that were generated against peptide immunogens prepared by maleimide conjugation; Immobilize antibodies in oriented manner through hinge-region sulfhydryls to ensure that antigen binding sites are not sterically hindered for antigen affinity purification
Specifications
Specifications
| Content And Storage | Upon receipt store product in the dark at 4°C. Product is shipped at ambient temperature. |
| Form | Liquid Suspension |
| Product Line | SulfoLink |
| Quantity | 10 mL |
| Type | Resin |
| Stationary Phase | Haloacetyl |
| Sufficient For | 1 mg Peptide or 10 mg Antibody per mL of Resin |
| Format | Bottle |
| Column Type | Affinity, Agarose Resin |
Frequently Asked Questions (FAQs)
SulfoLink Resin can bind approximately 5 mg of reduced IgG per mL of support. For peptides, the capacity is approximately 1 mg of peptide per mL of gel. Capacity for other compounds will vary.
SulfoLink Immobilization is compatible with 3-4 M fresh urea or guanidine. Alternatively, dissolve the peptide in 100% DMSO. Add the peptide in DMSO to the coupling buffer so that the DMSO does not exceed 20% of the final solution.
No. The resin will no longer react with subsequent sulfhydryls after the ligand is attached and the remaining active sites are blocked.
The columns can be reused at least 10 times without loss of activity.
SulfoLink Resin is crosslinked 6% beaded agarose that has been derivatized with a 12-atom spacer arm that ends in an iodoacetyl group. This spacer arm is helpful for peptides or large proteins that might otherwise be sterically hindered in binding their ligand. The iodoacetyl group reacts with free sulfhydryls at pH 7.5-8.5 to form covalent thioether bonds. The sulfhydryl must be present in the reduced form because disulfides will not react with the iodoacetyl group.
For Research Use Only. Not for use in diagnostic procedures.