missing translation for 'onlineSavingsMsg'
Learn More

Sulfo Orange, 0.04% (w/v) Indicator Solution, Spectrum™ Chemical

Catalog No. 18614181
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614181 100 mL Poly Bottle
18614182 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18614181 Supplier Spectrum Chemical Mfg Cor Supplier No. S595100ML

Chemical Identifiers

CAS 547-57-9 , 7732-18-5 , 1310-73-2

Specifications

Grade Chemical Solution
Packaging Poly Bottle
Color Orange to Red
Quantity 100 mL
Chemical Name or Material Sulfo Orange

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.