missing translation for 'onlineSavingsMsg'
Learn More

Sulfide IC Standard, MilliporeSigma™ Supelco™

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

$160.30 - $333.42

Specifications

Concentration 1000 μg/mL in 1% NaOH
Type Analytical Standard
Grade Analytical Standard
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111004779
View Documents
Merck Emd Millipore
ICS023100ML
100 mL
Each for $160.30
Only null left
 
111004394
View Documents
Merck Emd Millipore
ICS023500ML
500 mL
Each for $333.42
Only null left
 
Description

Description

Featured Industries: Environmental
Specifications

Specifications

1000 μg/mL in 1% NaOH
Analytical Standard
Analytical Standard
Videos
SDS
Documents

Documents