missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfathiazole, ≥98.0%, MilliporeSigma™ Supelco™
Sulfathiazole is a broad-spectrum antibacterial drug, belonging to the class of sulfonamides, used in veterinary and human medication for the treatment of infections. It is also involved in promoting growth of livestock and fish.
$147.86 - $147.86
Specifications
| Packaging | Poly Bottle |
|---|---|
| CAS | 72-14-0 |
| Physical Form | Neat |
| Linear Formula | C9H9N3O2S2 |
| MDL Number | MFCD00005319 |
Description
Environmental and Pharmaceutical (small molecule)Specifications
| Poly Bottle | |
| Neat | |
| MFCD00005319 | |
| 2-Sulfanilamidothiazole; 4-Amino-N-(2-thiazolyl)benzenesulfonamide; N1-(2-Thiazolyl)sulfanilamide | |
| 255.32 | |
| Analytical Standard |
| 72-14-0 | |
| C9H9N3O2S2 | |
| 200°C to 202°C | |
| Analytical Standard | |
| ≥98.0% |