missing translation for 'onlineSavingsMsg'
Learn More

Sulfate Standard, Certified, 1000ppm ±10ppm (1mL = 1mg SO4), LabChem™

Catalog No. LC255101
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC255101 500 mL Poly Bottle
LC255102 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC255101 Supplier LabChem Supplier No. LC255101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7757-82-6 , 7732-18-5
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Packaging Poly Bottle
CAS Max % 99.85, 0.15
Identification Passes Test
Viscosity 1.35cSt
Physical Form Liquid
Concentration 1000 ppm ±10 ppm (1mL = 1mg SO4)
Linear Formula Na2SO4
Color Colorless
Solubility Soluble in water
Quantity 500 mL
Formula Weight 142.04
Grade Certified
Chemical Name or Material Sulfate Standard, 1000ppm (1mL = 1mg SO4)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature