missing translation for 'onlineSavingsMsg'
Learn More

Sulfate Standard, 1000 ppm SO42-, Ricca Chemical

Catalog No. 81124
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
81124 120 mL Natural Poly Bottle
811216 500 mL Natural Poly Bottle
811232 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 81124 Supplier Ricca Chemical Company Supplier No. 81124

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7757-82-6 , 7732-18-5
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 1000ppm
Equivalence 1mL = 1mg SO42-
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 120 mL
Chemical Name or Material Sulfate Standard