missing translation for 'onlineSavingsMsg'
Learn More

Sulfasalazine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 461241000

Quantity 100 g

 View more versions of this product

Catalog No. AC461241000


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

599-79-1
398.39
(3Z)-6-oxo-3-(2-{4-[(pyridin-2-yl)sulfamoyl]phenyl}hydrazin-1-ylidene)cyclohexa-1,4-diene-1-carboxylic acid
C18H14N4O5S
OQANPHBRHBJGNZ-BKUYFWCQSA-N
OC(=O)C1=C\C(C=CC1=O)=N/NC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=N1

Specifications

Sulfasalazine
96.0 % min. (HPLC)
100 g
OQANPHBRHBJGNZ-BKUYFWCQSA-N
(3Z)-6-oxo-3-(2-{4-[(pyridin-2-yl)sulfamoyl]phenyl}hydrazin-1-ylidene)cyclohexa-1,4-diene-1-carboxylic acid
398.39
599-79-1
C18H14N4O5S
Solubility aq. soln.: insoluble. Other solubilities: slightly soluble in alcohols, insoluble in ether, benzene, chloroform
OC(=O)C1=C\C(C=CC1=O)=N/NC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=N1
398.39
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement May cause an allergic skin reaction.

May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Suspected of causing cancer.

GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

In case of inadequate ventilation wear respiratory protection.

IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.

If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Danger

EINECSNumber : 209-974-3

SDS
Documents

Documents