missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilic Acid, Anhydrous, ACS, 98-102%, Spectrum™ Chemical

Catalog No. p-7166414
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614172 25 g Poly Bottle
18614168 125 g Poly Bottle
18614170 500 g Poly Bottle
18614169 12 kg Poly Bottle
18614171 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18614172 Supplier Spectrum Chemical Mfg Cor Supplier No. S170325GM
Only null left

Chemical Identifiers

CAS 121-57-3
Molecular Formula C6H7NO3S
Molecular Weight (g/mol) 173.19
InChI Key HVBSAKJJOYLTQU-UHFFFAOYSA-N
IUPAC Name 4-aminobenzene-1-sulfonic acid
SMILES NC1=CC=C(C=C1)S(O)(=O)=O

Specifications

CAS Max % 1
Boiling Point 300°C
Ignition Residue 0.0001
Packaging Poly Bottle
Quantity 25 g
Percent Purity 98 to 102%
Grade ACS
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.