missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilamide 99.0+%, TCI America™

Catalog No. p-7135821
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S011925G 25 g
S0119100G 100 g
S0119500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S011925G Supplier TCI America Supplier No. S011925G

Chemical Identifiers

CAS 63-74-1
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
MDL Number MFCD00007939
InChI Key FDDDEECHVMSUSB-UHFFFAOYSA-N
Synonym sulfanilamide, sulphanilamide, sulfamine, sulphonamide, p-aminobenzenesulfonamide, sulfonylamide, sulfonamide, p-aminobenzenesulfamide, bacteramid, streptasol
PubChem CID 5333
ChEBI CHEBI:45373
IUPAC Name 4-aminobenzenesulfonamide
SMILES C1=CC(=CC=C1N)S(=O)(=O)N

Specifications

Melting Point 166°C
Color White
pH 5.8 to 6.1 (0.5% H2O soln.)
Quantity 25 g
Formula Weight 172.20
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sulfanilamide
EINECSNumber (3)-2179&(3)-1973
RTECSNumber WO8400000
TSCA Yes