missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilamide, 98%

Catalog No. AAA1300122
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1300122 100 g
AAA1300136 500 g
AAA130010E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1300122 Supplier Thermo Scientific Chemicals Supplier No. A1300122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 63-74-1 | C6H8N2O2S | 172.202 g/mol

Sulfonamide antibacterial Sulfanilamide is an antibacterial and used in the treatment of vaginal yeast infections. It is a competitive inhibitor of dihydropteroate synthestase to block the synthesis of folic acid. It serves as an intermediate for the preparation of 2,6-disubstituted anilines by electrophilic substitution followed by removal of the sulfonamide blocking group by desulfonation with sulfuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfonamide antibacterial Sulfanilamide is an antibacterial and used in the treatment of vaginal yeast infections. It is a competitive inhibitor of dihydropteroate synthestase to block the synthesis of folic acid. It serves as an intermediate for the preparation of 2,6-disubstituted anilines by electrophilic substitution followed by removal of the sulfonamide blocking group by desulfonation with sulfuric acid.

Solubility
Soluble in water, acetone, ethanol, glycerol, propylene glycol and hydrochloric acid. Insoluble in chloroform, ether, benzene and petroleum ether.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 63-74-1
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
MDL Number MFCD00007939
InChI Key FDDDEECHVMSUSB-UHFFFAOYSA-N
Synonym sulfanilamide, sulphanilamide, sulfamine, sulphonamide, p-aminobenzenesulfonamide, sulfonylamide, sulfonamide, p-aminobenzenesulfamide, bacteramid, streptasol
PubChem CID 5333
ChEBI CHEBI:45373
IUPAC Name 4-aminobenzenesulfonamide
SMILES C1=CC(=CC=C1N)S(=O)(=O)N

Specifications

Melting Point 163°C to 168°C
Density 1.08
Quantity 100 g
Beilstein 511852
Merck Index 14,8925
Solubility Information Soluble in water,acetone,ethanol,glycerol,propylene glycol and hydrochloric acid. Insoluble in chloroform,ether,benzene and petroleum ether.
Formula Weight 172.21
Percent Purity 98%
Chemical Name or Material Sulfanilamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-563-4
RTECSNumber WO8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only